Learn More
Chemetrics™ Chlorine Dioxide CHEMets™ Refill
Product Code. 11761883
Change view
Click to view available options
No. of Tests:
30
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests Unit Size
11761883 30 Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11761883 Supplier Chemetrics™ Supplier No. R2705
  • ONLINE EXCLUSIVE PRICE
    31.00€
    29.56€ / Pack of 30
    Save 1.44€
    4% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Achieve optimal oxidation with chlorine dioxide. Chemetrics™ Chlorine Dioxide — CHEMets™ Refill is used as an oxidizing microbiocide in industrial water-cooling treatment. It is used as a bleaching agent in the pulp and paper industry.
  • Oxidizing microbiocide
  • Used as a bleaching agent in the pulp industry
  • Disinfectant in municipal water treatment
Specifications
No. of Tests 30
Test Method DPD