missing translation for 'onlineSavingsMsg'
Learn More

Chlormequat chloride, Dr. Ehrenstorfer

Product Code. 30313905
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313905 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313905 Supplier Dr. Ehrenstorfer Supplier No. DRECA11340000100MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlormequat chloride
CAS 999-81-5
Analyte or Component Names Chlormequat chloride
Shipping Condition +20°C
Molecular Formula C5 H13 Cl N . Cl
Certifications/Compliance ISO 17034
Synonym Ethanaminium, 2-chloro-N,N,N-trimethyl-, chloride (1:1), (2-Chloroethyl)trimethylammonium chloride (6CI,7CI), Ammonium, (2-chloroethyl)trimethyl-, chloride (8CI), Ethanaminium, 2-chloro-N,N,N-trimethyl-, chloride (9CI), (β-Chloroethyl)trimethylammonium chloride, 60CS16, Antywylegacz, Antywylegacz Plynny, Antywylegacz plynny 675SL, Barleyquat B, Bercema CCC, CCC, Chlorcholine chloride, Chlormequat chloride, Chlorocholine chloride, Cycocel, Cycocel Extra, Cycogan, Cysocel, Halloween, Lihocin, Microcil, New 5C Cycocel, New '5c' Cycocel, Retacel, Stabilan, TUR, Trimethyl-β-chloroethylammonium chloride, Tuval, WR 62, Zar, Chlormequat-chloride
InChI Formula InChI=1S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].C[N+](C)(C)CCCl
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 158.07
Quantity 100 mg
Formula Weight 157.0425 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.