missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic Acid-13C2, TRC

Product Code. 30006618
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006618 100 mg 100mg
18072720 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006618 Supplier Toronto Research Chemicals Supplier No. TRCC363367100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1839-15-2
Molecular Formula 13C2H3ClO2
Molecular Weight (g/mol) 96.48
Synonym Monochloroacetic Acid-13C2, NSC 142-13C2, NSC 42970-13C2;
IUPAC Name 2-chloroacetic acid
SMILES O[13C]([13C]([H])([H])Cl)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)/i1+1,2+1
Formula Weight 95.9889
Percent Purity >95%
Chemical Name or Material Chloroacetic Acid-13C2
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.