missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene-4-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Chlorobenzene-4-D |
| CAS | 13122-34-4 |
| Molecular Formula | C6 D H4 Cl |
| Quantity | 0.25 g |
| Synonym | Benzene-d, 4-chloro- (9 CI, ACI), 4-Chlorobenzene-d (ACI), Benzene-d, p-chloro- (7 CI, 8 CI), Chlorobenzene-4-d, Chlorobenzene-4-D |
| InChI Formula | InChI=1 S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5 H/i1D |
| SMILES | [2 H]c1ccc(Cl)cc1 |
| IUPAC Name | 1-chloro-4-deuteriobenzene |
| Molecular Weight (g/mol) | 113.5631 |
| Formula Weight | 113.0143 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction