missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(4-methoxyphenyl)phosphine, 98+%

Product Code. 11475630
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475630 1 g 1g
11485630 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475630 Supplier Thermo Scientific Alfa Aesar Supplier No. H25887.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13685-30-8 | C14H14ClO2P | 280.69 g/mol

Chlorobis(4-methoxyphenyl)phosphine is used an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(4-methoxyphenyl)phosphine is used an pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13685-30-8
Molecular Formula C14H14ClO2P
Molecular Weight (g/mol) 280.69
MDL Number MFCD04972301
InChI Key YTFQUBRFOJIJOZ-UHFFFAOYSA-N
Synonym bis 4-methoxyphenyl chlorophosphine, chlorobis 4-methoxyphenyl phosphine, chlorobis 4-methoxyphenyl phosphane, chloro-bis 4-methoxyphenyl phosphane, acmc-20alng, p,p-bis 4-methoxyphenyl phosphinous chloride, phosphinous chloride,p,p-bis 4-methoxyphenyl
PubChem CID 4379930
SMILES COC1=CC=C(C=C1)P(Cl)C1=CC=C(OC)C=C1

Specifications

Melting Point 61°C to 64°C
Boiling Point 140°C (1.5 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name chlorobis(4-methoxyphenyl)phosphane
Formula Weight 280.69
Percent Purity ≥98%
Chemical Name or Material Chlorobis(4-methoxyphenyl)phosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.