missing translation for 'onlineSavingsMsg'
Learn More

Chlorocarbonylsulfenyl Chloride, TRC

Product Code. 30362441
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362441 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362441 Supplier Toronto Research Chemicals Supplier No. TRCC3684335G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorocarbonylsulfenyl Chloride
CAS 2757-23-5
Molecular Formula CCl2OS
Quantity 5 g
Synonym (Chlorocarbonyl)sulfenyl Chloride, (Chloroformyl)sulfur Chloride, (Chlorothio)formyl Chloride, 1-(Chlorothio)formyl Chloride, Chloroformylsulfenyl Chloride, Chlorosulfenylcarbonyl Chloride
InChI Formula InChI=1S/CCl2OS/c2-1(4)5-3
SMILES ClSC(=O)Cl
IUPAC Name S-chloro chloromethanethioate
Molecular Weight (g/mol) 130.98
Formula Weight 129.9047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.