Learn More
Chlorocitalopram, Hydrobromide, TRC
Product Code. 30003430
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003430 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30003430 Supplier Toronto Research Chemicals Supplier No. TRCC3650901MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 64169-58-0
Molecular Formula C20 H21 Cl N2 O . Br H
Quantity 1 mg
InChI Formula InChI=1S/C20H21ClN2O.BrH/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;/h4-9,12H,3,10-11,14H2,1-2H3;1H
SMILES Br.CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccc(Cl)cc3
IUPAC Name 1-(4-chlorophenyl)-1-[3-(dimethylamino)propyl]-3H-2-benzofuran-5-carbonitrile;hydrobromide
Molecular Weight (g/mol) 421.76
Formula Weight 420.0604
Chemical Name or Material Chlorocitalopram, Hydrobromide