missing translation for 'onlineSavingsMsg'
Learn More

Chloromethanesulfonyl Chloride, TRC

Product Code. 30367135
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367135 Supplier Toronto Research Chemicals Supplier No. TRCC4276991G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloromethanesulfonyl Chloride
CAS 3518-65-8
Molecular Formula CH2Cl2O2S
Quantity 1 g
Synonym 1-Chloromethanesulfonyl Chloride, (Chloromethyl)sulfonyl Chloride
InChI Formula InChI=1S/CH2Cl2O2S/c2-1-6(3,4)5/h1H2
SMILES ClCS(=O)(=O)Cl
IUPAC Name chloromethanesulfonyl chloride
Molecular Weight (g/mol) 149
Formula Weight 147.9153
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.