Learn More
(Chloromethyl)cyclopropane, 97%
Product Code. 11357076
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357076 5 g 5g
11367076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11357076 Supplier Thermo Scientific Alfa Aesar Supplier No. L06986.06
Request Bulk or Custom Format
CAS: 5911-08-0 | C4H7Cl | 90.55 g/mol
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4?-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5911-08-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00001307
InChI Key ZVTQWXCKQTUVPY-UHFFFAOYSA-N
Synonym chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane
PubChem CID 80013
SMILES ClCC1CC1
Specifications
Melting Point -91°C
Density 0.98
Boiling Point 87°C to 89°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.436
Quantity 5 g
UN Number UN1993
Beilstein 635697
Solubility Information Not miscible in water.
IUPAC Name chloromethylcyclopropane
Formula Weight 90.55
Percent Purity 97%
Chemical Name or Material (Chloromethyl)cyclopropane