missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)trichlorosilane, 97%

Product Code. 11329165
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329165 5 g 5g
11339165 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329165 Supplier Thermo Scientific Alfa Aesar Supplier No. L16433.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 1558-25-4 | CH2Cl4Si | 183.912 g/mol

(chloromethyl)trichlorosilane is usede in the synthesis of a highly branched polycarbosilane. Ferrocenophane has been prepared from (chloromethyl)trichlorosilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(chloromethyl)trichlorosilane is usede in the synthesis of a highly branched polycarbosilane. Ferrocenophane has been prepared from (chloromethyl)trichlorosilane.

Solubility
Reacts with water.

Notes
Air & moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, air and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1558-25-4
Molecular Formula CH2Cl4Si
Molecular Weight (g/mol) 183.912
MDL Number MFCD00000873
InChI Key FYTPGBJPTDQJCG-UHFFFAOYSA-N
Synonym chloromethyl trichlorosilane, trichloro chloromethyl silane, silane, trichloro chloromethyl, chloromethyltrichlorosilane, silane, chloromethyl trichloro, unii-03tzm169kh, silane,trichloro chloromethyl, silane cm, cl3sich2cl, acmc-20apm5
PubChem CID 15258
IUPAC Name trichloro(chloromethyl)silane
SMILES C([Si](Cl)(Cl)Cl)Cl

Specifications

Boiling Point 117°C to 118°C
Quantity 5 g
Linear Formula ClCH2SiCl3
UN Number UN3390
Beilstein 1811640
Flash Point 69°C (156°F)
Formula Weight 183.93
Percent Purity 97%
Odor Acrid
Chemical Name or Material (Chloromethyl)trichlorosilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.