missing translation for 'onlineSavingsMsg'
Learn More

Chloromethylisothiazolinone-methylisothiazolinone Mixture (~10% in water), TRC

Product Code. 30012351
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012351 10 g 10g
30004549 25 g 25g
30010186 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012351 Supplier Toronto Research Chemicals Supplier No. TRCC38734510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55965-84-9
Molecular Formula ((C4 H4 Cl N O S)c(C4 H5 N O S)c)mix
Molecular Weight (g/mol) 264.7523
Synonym 3(2H)-Isothiazolone, 5-chloro-2-methyl-, mixt. with 2-methyl-3(2H)-isothiazolone, 3(2H)-Isothiazolone, 2-methyl-, mixt. contg. (9CI), 5-Chloro-2-methyl-4-isothiazolin-3-one-2-methyl-4-isothiazolin-3-one mixt., 5-Chloro-2-methyl-4-isothiazolin-3-one-2-methyl-4-isothiazolin-3-one mixture, A 14, A 14 (antimicrobial), Acticide 14, Acticide 5008, Acticide LG, Acticide MV, Acticide MV 14, Acticide RS, Actocode MV, Alchinol D 15, Amerstat 252, BEK 500, Bio-Perge, Biocide K, Biocide K 10ME, Biocide K 10SG, Biocide SMB, Bodedex forte, Bodoxin, CI 15, CIT-MIT 14, CMIT-MIT mixt., Canguard CM 14, Canguard CM 1P5, Chloromethylisothiazolinone-methylisothiazolinone mixt., Fennosan IT 21, HY 606D, Hokucide R 150, Hydorol D 15, Isocil, Isocil (antibacterial), Isocil RW, Isothiazolinone chloride, JE 120, K 10SG, K 18500, KKM 43, KS 2, Kathon, Kathon 886, Kathon 886F, Kathon 886MW, Kathon 886W, Kathon CF 1400, Kathon CG, Kathon CG 110, Kathon CG/ICP, Kathon CG/ICP II, Kathon EDC, Kathon FP 1.5, Kathon ICP, Kathon LX, Kathon LX 1.5, Kathon LX 1400, Kathon LX Plus, Kathon LX-SF, Kathon LX-SF 25, Kathon LXE, Kathon MWC, Kathon PFM, Kathon RH 886, Kathon WT, Kathon biocide, Kordek MIX, Kuriverter EC 503, LH 2, LH 2 (corrosion inhibitor), LXE, Legend MK, M 30, M 30 (antibacterial), MBC 215, MV, Methylisothiazolinone-chloromethylisothiazolinone mixt., Methylchloroisothiazolinone-Methylisothiazolinone Mixture, Microcide III, Mirecide KW, Mirecide KW 600, Nalco 7330, Nipacide 15TR, Nipacide CI, Nipacide CI 15, Nipacide CI 15MV, Nipaguard CMB, Odycide B 330, Preventol D 7, ProClin 150, ProClin 200, ProClin 300, RHZL 03, RS, Rocima 625, SD 818, SD 818 (antibacterial), SS 411GY, Sharomix MCI, Slaoff 360, Somacide RS, Spectrus NX 1106, TST 25, Trelin CT, Tret-O-Lite XC 215, Yisaizuolintong, Zonen C, Zonen F, Zonen FP
IUPAC Name 5-chloro-2-methyl-isothiazol-3-one;2-methylisothiazol-3-one
SMILES CN1SC=CC1=O.CN2SC(=CC2=O)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H4ClNOS.C4H5NOS/c1-6-4(7)2-3(5)8-6;1-5-4(6)2-3-7-5/h2H,1H3;2-3H,1H3
Formula Weight 263.9794
Concentration or Composition (by Analyte or Components) Chloromethylisothiazolinone-methylisothiazolinone Mixture ~10%
Chemical Name or Material Chloromethylisothiazolinone-methylisothiazolinone Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.