missing translation for 'onlineSavingsMsg'
Learn More

Chloromethylphosphonothioic dichloride, 98%, Thermo Scientific™

Product Code. 11448377
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448377 Supplier Thermo Scientific Alfa Aesar Supplier No. A18456.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1983-27-3
Molecular Formula CH2Cl3PS
Molecular Weight (g/mol) 183.41
MDL Number MFCD00015876
InChI Key HRHKZXIMFUYRDS-UHFFFAOYSA-N
Synonym chloromethylphosphonothioic dichloride, chloromethylphosphonothioyl dichloride, acmc-1bqir, chloromethyl thiophosphoryl chloride, dichloro-chloromethyl-sulfanylidene, phosphonothioicdichloride, p-chloromethyl
PubChem CID 583951
SMILES ClCP(Cl)(Cl)=S

Specifications

Boiling Point 88°C to 90°C (30 mmHg)
Odor Acrid
Linear Formula ClCH2PSCl2
Refractive Index 1.575
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
IUPAC Name dichloro-(chloromethyl)-sulfanylidene-$l^{5}-phosphane
Formula Weight 183.42
Percent Purity 98%
Chemical Name or Material Chloromethylphosphonothioic dichloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.