missing translation for 'onlineSavingsMsg'
Learn More

Chloropentafluorobenzene, 98+%

Product Code. 11418336
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418336 50 g 50g
11408336 10 g 10g
11428336 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418336 Supplier Thermo Scientific Alfa Aesar Supplier No. A15384.18
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 344-07-0 | C6ClF5 | 202.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-07-0
Molecular Formula C6ClF5
Molecular Weight (g/mol) 202.51
MDL Number MFCD00000534
InChI Key KGCDGLXSBHJAHZ-UHFFFAOYSA-N
Synonym chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
PubChem CID 9579
IUPAC Name 1-chloro-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Cl)C(F)=C1F

Specifications

Density 1.568
Boiling Point 116°C to 118°C
Refractive Index 1.424
Quantity 50 g
Beilstein 1819389
Formula Weight 202.51
Percent Purity ≥98%
Chemical Name or Material Chloropentafluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.