missing translation for 'onlineSavingsMsg'
Learn More

Chloroxuron, TRC

Product Code. 30011527
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011527 500 mg 500mg
30004236 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011527 Supplier Toronto Research Chemicals Supplier No. TRCC427953500MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1982-47-4
Molecular Formula C15 H15 Cl N2 O2
Molecular Weight (g/mol) 290.74
Synonym Urea, 3-[p-(p-chlorophenoxy)phenyl]-1,1-dimethyl- (7CI,8CI), N'-[4-(4-Chlorophenoxy)phenyl]-N,N-dimethylurea, 3-[4-(p-Chlorophenoxy)phenyl]-1,1-dimethylurea, 3-[p-(p-Chlorophenoxy)phenyl]-1,1-dimethylurea, C 1983, Chloroxifenidim, Chloroxuron, Gesamoos, N-4-(p-Chlorophenoxy)phenyl-N',N'-dimethyl urea, N1-[4-(4-Chlorophenoxy)phenyl]-N,N-dimethylurea, Norex, Tenoran
IUPAC Name 3-[4-(4-chlorophenoxy)phenyl]-1,1-dimethylurea
SMILES CN(C)C(=O)Nc1ccc(Oc2ccc(Cl)cc2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H15ClN2O2/c1-18(2)15(19)17-12-5-9-14(10-6-12)20-13-7-3-11(16)4-8-13/h3-10H,1-2H3,(H,17,19)
Formula Weight 290.0822
Chemical Name or Material Chloroxuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.