missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine EP Impurity D (as Hydrochloride), Mikromol™

Product Code. 18053500
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18053500 25 mg 25mg
18042710 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18053500 Supplier Mikromol Supplier No. MM0420.070025
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Chlorpromazine EP Impurity D (as Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethylchlorpromazine Hydrochloride
CAS 3953-65-9
Analyte or Component Names Desmethylchlorpromazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H17 Cl N2 S . Cl H
Certifications/Compliance ISO 17025
Synonym 3-(2-Chloro-10H-phenothiazin-10-yl)-N-methylpropan-1-amine Hydrochloride (Desmethylchlorpromazine Hydrochloride), Chlorpromazine Hydrochloride Imp. D (EP), Chlorpromazine Imp. D (EP), 3-(2-Chloro-10H-phenothiazin-10-yl)-N-methylpropan-1-amine Hydrochloride, Chlorpromazine Hydrochloride Imp. D (EP) as Hydrochloride, Desmethylchlorpromazine Hydrochloride, Chlorpromazine Hydrochloride Impurity D as Hydrochloride, Chlorpromazine Impurity D as Hydrochloride
InChI Formula InChI=1S/C16H17ClN2S.ClH/c1-18-9-4-10-19-13-5-2-3-6-15(13)20-16-8-7-12(17)11-14(16)19;/h2-3,5-8,11,18H,4,9-10H2,1H3;1H
SMILES Cl.CNCCCN1c2ccccc2Sc3ccc(Cl)cc13
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N-methylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 341.3
Quantity 25 mg
Formula Weight 340.0568 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.