missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine Hydrochloride, TRC

Product Code. 30356776
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356776 1 g 1g
30362271 10 g 10g
30354100 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30356776 Supplier Toronto Research Chemicals Supplier No. TRCC4247501G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-09-0
Molecular Formula C17 H19 Cl N2 S . Cl H
Molecular Weight (g/mol) 355.33
Synonym 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, monohydrochloride (9CI), Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, monohydrochloride (8CI), 10-(3-Dimethylaminopropyl)-2-chlorophenothiazine monohydrochloride, 2-Chloro-10-(3-dimethylaminopropyl)phenothiazine monohydrochloride, 3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1-amine hydrochloride, Aminazin hydrochloride, Aminazin monohydrochloride, Ampliactil monohydrochloride, CPZ, Chloractil, Chlorazin, Chloropromazine hydrochloride, Chloropromazine monohydrochloride, Chlorpromazine hydrochloride, Chlorpromazine monohydrochloride, Chlorpromazinium chloride, Contomin, Hebanil, Hibanil, Hibernal, Klorproman, Klorpromex, Largactil, Largactil monohydrochloride, Largaktyl, Marazine, Megatil, Neurazine, Norcozine, Plegomazin, Promacid, Promapar, Propaphen, Propaphenin hydrochloride, Sonazine, Taroctyl, Thorazine hydrochloride, Torazina, Tranzene
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H
Formula Weight 354.0724
Percent Purity >95%
Chemical Name or Material Chlorpromazine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.