missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine, TRC

Product Code. 18013520
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18013520 50 mg 50mg
18081810 500 mg 500mg
18001530 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18013520 Supplier Toronto Research Chemicals Supplier No. TRCC42476850MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-53-3
Molecular Formula C17 H19 Cl N2 S
Molecular Weight (g/mol) 318.86
Synonym 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]- (7CI,8CI), 2-Chloro-N,N-dimethyl-10H-phenothiazine-10-propanamine, 2-Chloro-10-[3-(dimethylamino)propyl]phenothiazine, 2-Chloropromazine, 2601A, Aminazin, Aminazine, Ampliactil, Amplictil, BC 135, CPZ, Chlor-Promanyl, Chlordelazin, Chlorderazin, Chlorpromados, Chlorpromazine, Chlropromados, Elmarin, Esmind, Fenactil, Fenaktyl, Fraction AB, HL 5746, Largactilothiazine, Largactyl, Largaltil, Megaphen, NSC 167745, NSC 226514, Noiafren, Novomazina, Phenactyl, Proma, Promactil, Promazil, Propaphenin, Prozil, RP 4560, Relitil, SKF 2601A, Sanopron, Sedatil, Thorazin, Thorazine, Wintermin
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H19ClN2S/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20/h3-4,6-9,12H,5,10-11H2,1-2H3
Formula Weight 318.0957
Percent Purity >95%
Chemical Name or Material Chlorpromazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.