missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide EP Impurity B, Mikromol™

Product Code. 17949604
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949604 100 mg 100mg
17939304 25 mg 25mg
30461308 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17949604 Supplier Mikromol Supplier No. MM1635.01
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Chlorpropamide EP Impurity B
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dipropylurea
CAS 623-95-0
Analyte or Component Names 1,3-Dipropylurea
Shipping Condition Room Temperature
Molecular Formula C7 H16 N2 O
Certifications/Compliance ISO 17025
Synonym N,N'-Dipropylurea, Urea, 1,3-dipropyl- (6CI,7CI,8CI), 1,3-Di-n-propylurea, 1,3-Dipropylurea, N,N'-Dipropylurea, Chlorpropamide Impurity B
InChI Formula InChI=1S/C7H16N2O/c1-3-5-8-7(10)9-6-4-2/h3-6H2,1-2H3,(H2,8,9,10)
SMILES CCCNC(=O)NCCC
IUPAC Name 1,3-dipropylurea
Molecular Weight (g/mol) 144.215
Quantity 100 mg
Formula Weight 144.126 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.