missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide, Thermo Scientific™

Increases insulin secretion

Brand:  Thermo Scientific Alfa Aesar J64110.14

Quantity 25g

 View more versions of this product

Product Code. 15466209

  • 72.70€ / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

Solubility
Soluble in ethanol (1:12), acetone (1:5), chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

94-20-2
276.735
RKWGIWYCVPQPMF-UHFFFAOYSA-N
2727
1-(4-chlorophenyl)sulfonyl-3-propylurea
C10H13ClN2O3S
MFCD00079004
chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
CHEBI:3650
CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

94-20-2
MFCD00079004
14,2186
Soluble in ethanol (1:12),acetone (1:5),chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.
CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl
276.735
CHEBI:3650
Chlorpropamide
C10H13ClN2O3S
25g
chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
RKWGIWYCVPQPMF-UHFFFAOYSA-N
1-(4-chlorophenyl)sulfonyl-3-propylurea
2727
276.74
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.