missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide, Thermo Scientific™

Increases insulin secretion

72.70€ - 233.00€

Chemical Identifiers

CAS 94-20-2
Molecular Formula C10H13ClN2O3S
Molecular Weight (g/mol) 276.735
MDL Number MFCD00079004
InChI Key RKWGIWYCVPQPMF-UHFFFAOYSA-N
Synonym chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
PubChem CID 2727
ChEBI CHEBI:3650
IUPAC Name 1-(4-chlorophenyl)sulfonyl-3-propylurea
SMILES CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15466209
SDS View Documents
Thermo Scientific Alfa Aesar
J64110.14
25g
72.70€
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15476209
SDS View Documents
Thermo Scientific Alfa Aesar
J64110.22
233.00€
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

Solubility
Soluble in ethanol (1:12), acetone (1:5), chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

94-20-2
276.735
RKWGIWYCVPQPMF-UHFFFAOYSA-N
2727
1-(4-chlorophenyl)sulfonyl-3-propylurea
C10H13ClN2O3S
MFCD00079004
chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
CHEBI:3650
CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

94-20-2
MFCD00079004
chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
RKWGIWYCVPQPMF-UHFFFAOYSA-N
1-(4-chlorophenyl)sulfonyl-3-propylurea
2727
276.74
C10H13ClN2O3S
14,2186
Soluble in ethanol (1:12),acetone (1:5),chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.
CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl
276.735
CHEBI:3650
Chlorpropamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

P201-P202-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c

H312+H332-H351

missing translation for 'einecsNumber' : 202-314-5

missing translation for 'rtecsNumber' : YS6650000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.