missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropham, TRC

Product Code. 17987399
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987399 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17987399 Supplier Toronto Research Chemicals Supplier No. TRCC424905250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 101-21-3
Molecular Formula C10 H12 Cl N O2
Quantity 250 mg
Synonym Carbamic acid, (3-chlorophenyl)-, 1-methylethyl ester (9CI), Carbanilic acid, m-chloro-, isopropyl ester (6CI,7CI,8CI), Bud Nip, Bygran, CI-IPC, CIPC, Chlor IFC, Chlor IFK, Chlor IPC, Chloro-IPC, Chloropropham, Chlorpropham, Cl-IFK, Elbanil, Furloe, Furloe 3EC, IPCPC, Isopropyl 3-chlorocarbanilate, Isopropyl N-(3-chlorophenyl)carbamate, Isopropyl N-(m-chlorophenyl)carbamate, Isopropyl m-chlorocarbanilate, Keim-Stop, Liro CIPC, Metoxon, Mirvale, NSC 29466, Neo Stop 5, Nexoval, Preventol, Preventol 56, Shield Potato Sprout Inhibitor, Triherbicide CIPC, Y 3, m-Chlorocarbanilic acid isopropyl ester
InChI Formula InChI=1S/C10H12ClNO2/c1-7(2)14-10(13)12-9-5-3-4-8(11)6-9/h3-7H,1-2H3,(H,12,13)
SMILES CC(C)OC(=O)Nc1cccc(Cl)c1
IUPAC Name propan-2-yl N-(3-chlorophenyl)carbamate
Molecular Weight (g/mol) 213.66
Formula Weight 213.0557
Chemical Name or Material Chlorpropham
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.