Learn More
Invitrogen™ CHMP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530815
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (95%), Rat (96%), Chicken (81%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CHMP6 belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).
Specifications
| CHMP6 | |
| Polyclonal | |
| Unconjugated | |
| CHMP6 | |
| 2400004G01Rik; Charged multivesicular body protein 6; Chmp6; chromatin modifying protein 6; chromatin-modifying protein 6; hVps20; novel SNF7 protein; RGD1565325; vacuolar protein sorting-associated protein 20; VPS20 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96FZ7 | |
| CHMP6 | |
| Recombinant protein encompassing a sequence within the center region of human CHMP6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.