missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol, TRC

Product Code. 17988874
Change view
Click to view available options
Quantity:
50 mg
10 g
25 g
Unit Size:
10g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988874 25 g 25g
17969724 50 mg 50mg
17938204 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17988874 Supplier Toronto Research Chemicals Supplier No. TRCC43250125G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27 H46 O
Molecular Weight (g/mol) 386.65
Synonym Cholesterol (8CI), (3β)-Cholest-5-en-3-ol, (-)-Cholesterol, 3β-Hydroxycholest-5-ene, 5:6-Cholesten-3β-ol, Cholest-5-en-3β-ol, Cholesterin, Cholesteryl alcohol, Dythol, Lidinit, Lidinite, Marine Cholesterol, NSC 8798, Provitamin D, SyntheChol, δ5-Cholesten-3β-ol
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1
Formula Weight 386.3549
Percent Purity >95%
Chemical Name or Material Cholesterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.