missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid Sodium Salt, ≥98.5%, MP Biomedicals™

Product Code. 11426292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426292 1 kg 1kg
11416292 5 g 500g
11406292 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426292 Supplier MP Biomedicals Supplier No. 0210289701
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cholic Acid Sodium Salt, ≥98.5% is used in the solubilization of membrane-bound proteins in their native state.

  • Cholic Acid Sodium Salt, ≥98.5% is a component for the preparation of liposomes.
  • It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
  • Critical Micelle Concentration: 14 mM
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Formula Weight 430.6
Percent Purity ≥98%
Chemical Name or Material Cholic acid sodium salt

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.