missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid Sodium Salt, MP Biomedicals™

Product Code. 17891361 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17891361 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17891361 Supplier MP Biomedicals Supplier No. 02102897.3
  • ONLINE EXCLUSIVE PRICE
    59.55€
    56.70€ / 25g
    Save 2.85€
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used in the solubilization of membrane-bound proteins in their native state.

  • A component for the preparation of liposomes.
  • Used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
  • Biochemical solubilizing agent (a non-ionic, non-denaturing detergent).
  • Occurs inconjugation with glycine or taurine in bile of most vertebrates .
TRUSTED_SUSTAINABILITY

Specifications

CAS 361-09-1
Physical Form White Powder
pH 7.5 to 9.0 (2% aq. soln.)
Quantity 25 g
Molecular Formula C24H39NaO5
MDL Number MFCD00150749,MFCD00064138
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
Solubility Information Soluble in water (100mg/mL). Soluble in ethanol (40mg/mL, clear, colorless solution).
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Molecular Weight (g/mol) 430.56
PubChem CID 57349315
Formula Weight 430.6
Percent Purity ≥98.5%
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 14mM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.