missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline-d6 Chloride (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Choline-D6 (N,N-dimethyl-D6) Chloride |
| CAS | 3035513-80-2 |
| Molecular Formula | C5 D6 H8 N O . Cl |
| Quantity | 0.1 g |
| Synonym | Choline-D6 (N,N-dimethyl-D6) Chloride, Choline-d6 Chloride, 2-Hydroxy-N-methyl-N,N-di(methyl-d3)ethanaminium chloride (1:1), N,N-Bis(trideuteriomethyl)-N-(2-hydroxyethyl)-N-methylammonium chloride, Choline-d6 Chloride (N,N-dimethyl-d6) |
| InChI Formula | InChI=1 S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1 H/q+1;/p-1/i1D3,2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(CCO)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-hydroxyethyl-methyl-bis(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 145.6607 |
| Formula Weight | 145.1141 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction