missing translation for 'onlineSavingsMsg'
Learn More

Choline p-Toluenesulfonate Salt, TRC

Product Code. 17961965
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961965 5 g 5g
17910755 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17961965 Supplier Toronto Research Chemicals Supplier No. TRCC4330505G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55357-38-5
Molecular Formula C7 H7 O3 S . C5 H14 N O
Molecular Weight (g/mol) 275.36
Synonym 4-Methylbenzenesulfonate 2-Hydroxy-N,N,N-trimethylethanaminium, Choline Toluenesulfonate, Choline Tosylat
IUPAC Name 2-hydroxyethyl(trimethyl)ammonium;4-methylbenzenesulfonate
SMILES Cc1ccc(cc1)S(=O)(=O)[O-].C[N+](C)(C)CCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H8O3S.C5H14NO/c1-6-2-4-7(5-3-6)11(8,9)10;1-6(2,3)4-5-7/h2-5H,1H3,(H,8,9,10);7H,4-5H2,1-3H3/q;+1/p-1
Formula Weight 275.1191
Chemical Name or Material Choline p-Toluenesulfonate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.