Learn More
Chondroitin Sulfate Sodium Salt, TRC
Product Code. 30135998
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135998 5 g 5g
17958204 1 g 1g
17997914 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135998 Supplier Toronto Research Chemicals Supplier No. TRCC4327355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9082-07-9
Molecular Formula C28H40N2Na4O29S2
Synonym Chondroitin Hydrogen Sulfate Sodium Salt
IUPAC Name sodium (2S,3S,4R,5R,6R)-6-(((2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-((sulfonatooxy)methyl)tetrahydro-2H-pyran-4-yl)oxy)-3-(((2S,3R,4R,5R,6R)-3-acetamido-4-(((2R,3R,4S,5S,6S)-6-carboxylato-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-6-((sulfonatooxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4,5-dihydroxytetrahydro-2H-pyran-2-carboxylate
SMILES O[C@H]1[C@H](O[C@@H]([C@@H]2NC(C)=O)O[C@H](COS(=O)(O[Na])=O)[C@H](O)[C@@H]2O[C@@H]([C@@H]3O)O[C@H](C(O[Na])=O)[C@@H](O)[C@@H]3O)[C@@H](C(O[Na])=O)O[C@@H](O[C@H]4[C@@H](O)[C@@H](COS(=O)(O[Na])=O)O[C@@H](O)[C@@H]4NC(C)=O)[C@@H]1O
Specifications
Quantity 5 g
Chemical Name or Material Chondroitin sulfate sodium salt