missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHOP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15204-1-AP-20UL
This item is not returnable.
View return policy
Description
GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.Specifications
| CHOP | |
| Polyclonal | |
| Unconjugated | |
| DDIT3 | |
| CEBPZ, CHOP, CHOP 10, DDIT 3, DDIT3, GADD153 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13198, 1649, 29467 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35638, P35639, Q62857 | |
| DDIT3 | |
| CHOP Fusion Protein Ag7354 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction