Learn More
Christeyns™ Chriox Spray
Product Code. 17890245
Change view
Click to view available options
Size:
750 mL
5 L
Unit Size:
4.5 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17890245 750 mL 4.5 L
17800265 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17890245 Supplier Christeyns™ Supplier No. 391172
  • ONLINE EXCLUSIVE PRICE
    38.90€
    35.12€ / 4.5 L
    Save 3.78€
    9% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Chriox spray is an ecological, ready-to-use disinfectant based on peracetic acid.
  • Eco-detergent certified by ECOCERT Greenlife according to the ECOCERT standard
  • Suitable for cleaning all washable surfaces (including in the food industry)
  • Spray the pure product on the surface to be treated or preferably on a clean, nonwoven cloth, and leave on for 5 to 15 min.
Specifications
Product Type Surface Disinfectant
Color-coded No
Form Clear Liquid
pH 3 to 3.4
Scent Unscented
Sterility Non-sterile
Quantity 6 Bottles
Size 750 mL