Learn More
CHRM3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21978-1-AP-20UL
Description
The CHRM3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CHRM3. This antibody recognizes human, mouse, rat antigen. The CHRM3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CHRM3 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CHRM3, HM3 | |
| 20 μL | |
| 1131 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |