missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21379-1-AP-20UL
This item is not returnable.
View return policy
Description
Chromosome 15 encodes over 700 genes and comprises nearly 3% of the human genome. CHRFAM7A, a fusion protein comprised of FAM7A and AChR alpha7, is encoded by the CHRFAM7A gene, which is located on a region of human chromosome 15 that is associated with several neuropsychiatric disorders, such as schizophrenia and bipolar affective disorder. CHRFAM7A, also known as D-10 or CHRNA7, is a 412 amino acid multi-pass membrane protein that belongs to the nicotinic acetylcholine receptor (nAChR) subfamily of ligand-gated ion channel proteins, a protein superfamily that mediates signal transmission at synapses. CHRFAM7A is expressed in the hippocampus. Alternative splicing of the CHRFAM7A gene produces two splice variants.Specifications
| CHRNA7 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA7 | |
| CHRNA7, CHRNA7 2, NACHRA7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1139, 11441, 25302 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P36544, P49582, Q05941 | |
| Chrna7 | |
| CHRNA7 Fusion Protein Ag15980 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction