Learn More
Macherey-Nagel™ Chromafix™ HR-XCW Cartridge
Product Code. 12798252 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
12798252 155 mg Pack of 50
12757740 240 mg Pack of 50
12767740 500 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12798252 Supplier Macherey-Nagel™ Supplier No. 731774
  • ONLINE EXCLUSIVE PRICE
    254.39€
    243.60€ / Pack of 50
    Save 10.79€
    4% Off
View all promotions
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 155 mg
For Use With (Application) Basic Compounds Like Quaternary Amines, Basic Active Ingredients From Strongly Matrix-Contaminated Samples
Particle Size 45 or 85 μm
Sorbent Weak carboxylic acid modified polymeric cation exchanger
Type Chromafix HR-XCW Cartridge
Pore Size 50 to 60 Å
Quantity 50 Pack
Surface Area (Metric) 850 m2/g