missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromane-2-carboxylic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Chromane-2-carboxylic Acid |
| CAS | 51939-71-0 |
| Molecular Formula | C10 H10 O3 |
| Quantity | 10 g |
| Synonym | 2H-1-Benzopyran-2-carboxylic acid, 3,4-dihydro-, 2-Chromancarboxylic acid (6CI), 3,4-Dihydro-2H-1-benzopyran-2-carboxylic acid, (±)-2-Chromancarboxylic acid, 3,4-Dihydro-1-benzopyran-2-carboxylic acid, Chromane-2-carboxylic acid |
| InChI Formula | InChI=1S/C10H10O3/c11-10(12)9-6-5-7-3-1-2-4-8(7)13-9/h1-4,9H,5-6H2,(H,11,12) |
| SMILES | OC(=O)C1CCc2ccccc2O1 |
| IUPAC Name | 3,4-dihydro-2H-chromene-2-carboxylic acid |
| Molecular Weight (g/mol) | 178.18 |
| Formula Weight | 178.063 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction