Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and Beaded Rim
Product Code. 10693812
Change view
Click to view available options
Column Capacity:
15 mL
125 mL
1000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity Unit Size
10693812 1000 mL Each
10778461 125 mL Each
11984011 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10693812 Supplier Lenz Laborglasinstrumente™ Supplier No. 05445708
  • ONLINE EXCLUSIVE PRICE
    171.00€
    115.66€ / Pack of 1
    Save 55.34€
    32% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Description Chromatography Column
For Use With (Application) Chromatographic applications
Diameter (Metric) Inner 40 mm
Type Chromatography Column
Column Capacity 1000 mL
Length (Metric) 800 mm
Material Duran Glass
Diameter (Metric) Outer 40 mm
Includes PTFE Stopcock