Learn More
Hecht Karl™ Borosilicate Glass 3.3 Automatic Pipets
Click to view available options
Volume (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Includes | NS 29/32 Standard Ground Joint |
| Quantity | 1 Pack |
| Volume (Metric) | 10 mL |
| Type | Automatic Pipette |