Learn More
Metro™ Chrome Steel Shelves
Product Code. 11830300
Change view
Click to view available options
Dimensions (L x W):
1220 x 610 mm
1825 x 610 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11830300 1220 x 610 mm Each
11800320 1825 x 610 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11830300 Supplier Metro™ Supplier No. 5A557C
  • ONLINE EXCLUSIVE PRICE
    798.00€
    641.11€ / Pack of 1
    Save 156.89€
    19% Off
View all promotions
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Shelf
Material Chrome Steel
Diameter (Metric) 610 mm
Depth (Metric) 610 mm
Dimensions (L x W) 1220 x 610 mm
Dimensions (L x W x H) 1220 x 610 x 1895 mm
For Use With (Application) For storage in ventilated area, the air flow is not disturbed
Height (Metric) 189.5 cm
Length (Metric) 122 cm
Load Bearing Capacity (Metric) 270 kg
Shelf Type Complete Stationary Unit 5
Width (Metric) 610 mm