Learn More
Chromium Trioxide, ACS Reagent Grade, Ricca Chemical
Product Code. 31749323
Change view
Click to view available options
Quantity:
500 g
Packaging:
White Poly Wide Mouth Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31749323 500 g White Poly Wide Mouth Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31749323 Supplier Ricca Chemical Company Supplier No. RDCC0550500B1
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Specifications
CAS 1333-82-0
Color Purple to Red
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystalline Solid
Quantity 500 g
Molecular Formula CrO3
MDL Number MFCD00010952
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILES O=[Cr](=O)=O
IUPAC Name trioxochromium
Molecular Weight (g/mol) 99.99
PubChem CID 14915
ChEBI CHEBI:48240
Percent Purity ≥ 98%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Chromium Trioxide