missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Antibody (CHGA/1773R), Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NBP2-53140-20UG

 View more versions of this product

Product Code. 18287820

  • 352.00€ / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Chromogranin A Monoclonal specifically detects Chromogranin A in Human, Mouse, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chromogranin A
Monoclonal
0.2 mg/mL
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I
Rabbit
72 kDa
20 μg
Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers
1113
Store at 4C.
IgG κ
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
CHGA/1773R
Unconjugated
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
CHGA
Recombinant human full-length Chromogranin A protein
Protein A or G purified
RUO
Primary
Human, Rat (Negative)
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only