missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Rabbit, Clone: CHGA/1815R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 1113-RBM15-P0
Additional Details : Weight : 0.01000kg
Specifications
Chromogranin A | |
Finely granular cytoplasmic | |
CHGA/1815R | |
Unconjugated | |
NP_001266 | |
Rabbit | |
68-75kDa | |
Protein A/G Purified | |
RUO | |
1113 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
CHGA | |
Recombinant full-length human CHGA protein | |
PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |