missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carboxylic acid, 98%

Product Code. 11477057
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477057 1 g 1g
11487057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477057 Supplier Thermo Scientific Alfa Aesar Supplier No. A14422.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 39079-62-4 | C10H6O4 | 190.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39079-62-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00017338
InChI Key PCIITXGDSHXTSN-UHFFFAOYSA-N
Synonym chromone-3-carboxylic acid, 4-oxo-4h-chromene-3-carboxylic acid, 4-oxo-4h-1-benzopyran-3-carboxylic acid, maybridge1_005842, 3-carboxychromone, d04tgn, acmc-1ad65, 4-oxo-chromene-3-carboxylic acid, 4h-chromen-4-one-3-carboxylic acid
PubChem CID 181620
IUPAC Name 4-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C(=O)O

Specifications

Melting Point 198°C to 203°C
Quantity 1 g
Beilstein 1074111
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material Chromone-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.