Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose Kit
Product Code. 17363363
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17363363 20 Reactions 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17363363 Supplier Proteintech Supplier No. gtmak20
Kit including 20 reactions of GFP-Trap™ Magnetic Agarose and lysis, dilution, wash, and elution buffers.
  • The ChromoTek GFP-Trap™ Magnetic Agarose are affinity beads for IP of GFP-fusion proteins
  • Comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions