Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human IgG/Rabbit IgG, Fc-specific VHH, Biotinylated
Product Code. 17373463
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17373463 10 μL 10µL
17383463 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17373463 Supplier Proteintech Supplier No. shurbGB110
Nano-CaptureLigand for human IgG and rabbit IgG: Fc-specific single domain antibody/VHH [CTK0101], biotinylated.
  • The Nano-CaptureLigand™ human IgG captures and immobilizes the Fc-fragment of human IgG antibodies to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to human IgG antibodies with high affinity and in a site-directed manner
Specifications
Quantity 10 μL
Target Specificity Human IgE
Conjugate Biotin
Clonality Monoclonal
Content And Storage Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
For Use With (Application) BLI, SPR, ELISA