Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2a, Fc-specific VHH, Biotinylated
Description
- The Nano-CaptureLigand mouse IgG2a captures and immobilizes mouse IgG2a antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
- Consists of a biotinylated Nanobody that specifically binds to the mouse IgG2a subclass with high affinity and in a site-directed manner
Tekniske data
Tekniske data
| Quantity | 100 μL |
| Target Specificity | Fc-fragment of Mouse IgG2a |
| Conjugate | Biotin |
| Clonality | Monoclonal |
| Content And Storage | Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
| For Use With (Application) | BLI, SPR, ELISA |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?