Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Agarose Kit
Product Code. 17393453
Change view
Click to view available options
Quantity:
20 Reactions
Conditionnement:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
17393453 20 Reactions 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17393453 Fournisseur Proteintech Code fournisseur rtmak20
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Kit including 20 reactions of RFP-Trap™ Magnetic Agarose and lysis, dilution, wash, and elution buffers.
  • The ChromoTek RFP-Trap™ Magnetic Agarose are affinity beads for IP of RFP-fusion proteins
  • Comprises a RFP Nanobody/ VHH coupled to magnetic agarose
Spécification
Quantity 20 Reactions
Sufficient For 20 Reactions