Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Agarose Kit
Description
- Spot-Trap™ Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of Spot-tagged proteins
- Consists of an anti-Spot-Tag™ VHH/ Nanobody that is coupled to magnetic agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |