missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIB2 Antibody (09), DyLight 550 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05869R
This item is not returnable.
View return policy
Description
CIB2 Monoclonal antibody specifically detects CIB2 in Human samples. It is validated for ELISA, ELISA
Specifications
| CIB2 | |
| Monoclonal | |
| DyLight 550 | |
| 2810434I23Rik, calcium and integrin binding family member 2, DNA-dependent protein kinase catalytic subunit-interacting protein 2, Kinase-interacting protein 2, KIP 2, KIP2calcium and integrin-binding family member 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CIB2 (O75838; Met1-Ile187). | |
| 0.1 mL | |
| Cancer | |
| 10518 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 09 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction