missing translation for 'onlineSavingsMsg'
Learn More

Ciglitazone, TRC

Product Code. 30355424
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355424 1 mg 1mg
30355032 50 mg 50mg
30359799 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355424 Supplier Toronto Research Chemicals Supplier No. TRCC4410001MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74772-77-3
Molecular Formula C18 H23 N O3 S
Molecular Weight (g/mol) 333.45
IUPAC Name 5-[[4-[(1-methylcyclohexyl)methoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES CC1(COc2ccc(CC3SC(=O)NC3=O)cc2)CCCCC1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H23NO3S/c1-18(9-3-2-4-10-18)12-22-14-7-5-13(6-8-14)11-15-16(20)19-17(21)23-15/h5-8,15H,2-4,9-12H2,1H3,(H,19,20,21)
Formula Weight 333.1399
Chemical Name or Material Ciglitazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.