missing translation for 'onlineSavingsMsg'
Learn More

Cilazapril, TRC

Product Code. 30135985
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135985 50 mg 50mg
17961445 5 mg 5mg
17930835 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30135985 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC44120050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88768-40-5
Synonym (1S,9S)-9-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]octahydro-10-oxo-6H-pyridazino[1,2-a][1,2]diazepine-1-carboxylic Acid∣Ro 31-2848∣Vascace∣Yipingshu

Specifications

Format Neat
Molecular Formula C22H33N3O6
MDL Number MFCD00865788
Molecular Weight (g/mol) 417.5, 435.52
InChI Key JQRZBPFGBRIWSN-YOTVLOEGSA-N
SMILES O.CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@H]1CCCN2CCC[C@H](N2C1=O)C(O)=O
IUPAC Name (1S,9S)-9-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}-10-oxo-octahydro-1H-pyridazino[1,2-a][1,2]diazepine-1-carboxylic acid hydrate
Product Type API
Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.