missing translation for 'onlineSavingsMsg'
Learn More

Cilostazol, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18015842
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015842 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18015842 Supplier Tocris Bioscience Supplier No. 1692/10

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

PDE3A inhibitor. Also adenosine uptake inhibitor

Potent phosphodiesterase III A (PDE3A) inhibitor (IC50 = 0.2 μM) and inhibitor of adenosine uptake. Has antimitogenic, antithrombotic, vasodilatory and cardiotonic properties in vivo. Also affects lipid levels in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 73963-72-1
Molecular Formula C20H27N5O2
Quantity 10 mg
Synonym cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
InChI Key RRGUKTPIGVIEKM-UHFFFAOYSA-N
SMILES C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4
IUPAC Name 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 369.469
PubChem CID 2754
ChEBI CHEBI:31401
Formula Weight 369.47
Percent Purity >99%
Chemical Name or Material Cilostazol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.